missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonyl chloride, 99%, ACROS Organics™

Brand:  Acros Organics 173051000

Code : 29

Additional Details : CAS Number : 421-83-0 Weight : 0.15000kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 11964581

  • 596.00 € / 100g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

421-83-0
168.51
GRGCWBWNLSTIEN-UHFFFAOYSA-N
79000
FC(F)(F)S(Cl)(=O)=O
CClF3O2S
MFCD00007451
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
trifluoromethanesulfonyl chloride

Specifications

Trifluoromethanesulfonyl chloride
Authentic
1.3330 to 1.3350
29.0°C to 32.0°C
100g
98.5% min. (GC)
CF3SO2Cl
03,IV,35
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
GRGCWBWNLSTIEN-UHFFFAOYSA-N
trifluoromethanesulfonyl chloride
79000
Liquid
1.5800g/mL
Glass bottle
1.58
Colorless
421-83-0
CClF3O2S
MFCD00007451
09,485
Solubility in water: reacts
FC(F)(F)S(Cl)(=O)=O
168.51
168.52
99%
Videos
SDS Documents
Certificates