missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-4510484
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
421-83-0
CClF3O2S
168.51
MFCD00007451
GRGCWBWNLSTIEN-UHFFFAOYSA-N
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
79000
trifluoromethanesulfonyl chloride
FC(F)(F)S(Cl)(=O)=O
421-83-0
CClF3O2S
168.51
MFCD00007451
GRGCWBWNLSTIEN-UHFFFAOYSA-N
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
79000
trifluoromethanesulfonyl chloride
FC(F)(F)S(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.5800g/mL
Boiling Point 29.0°C to 32.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3SO2Cl
Refractive Index 1.3330 to 1.3350
Quantity 5 g
Beilstein 03,IV,35
Fieser 09,485
Specific Gravity 1.58
Solubility Information Solubility in water: reacts
Formula Weight 168.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'