missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, 98%

Product Code. 11925821 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11925821

Brand: Thermo Scientific Acros 214922500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6485-79-6 | C9H22Si | 158.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-79-6
Molecular Formula C9H22Si
Molecular Weight (g/mol) 158.36
MDL Number MFCD00009657
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
PubChem CID 6327611
IUPAC Name tri(propan-2-yl)silicon
SMILES CC(C)[SiH](C(C)C)C(C)C

Specifications

Density 0.7730g/mL
Boiling Point 84.0°C to 86.0°C (35.0 mmHg)
Flash Point 37°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]3SiH
Refractive Index 1.4340 to 1.4360
Quantity 250 g
Beilstein 04,IV,3907
Specific Gravity 0.773
Solubility Information Solubility in water: negligible
Formula Weight 158.36
Percent Purity 98%
Chemical Name or Material Triisopropylsilane, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triisopropylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.