missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-2799242
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
13170-43-9, 109-99-9, 108-88-3
C4H11ClMgSi
146.97
MFCD00009916
NAQATMJWCJCHOZ-UHFFFAOYSA-M
trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
2734900
C[Si](C)(C)C[Mg]Cl
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
13170-43-9, 109-99-9, 108-88-3
C4H11ClMgSi
146.97
MFCD00009916
NAQATMJWCJCHOZ-UHFFFAOYSA-M
trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
2734900
C[Si](C)(C)C[Mg]Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Green
CAS Min % 18.37, 80.22
CAS Max % 19.78, 81.63
Linear Formula (CH3)3Si-CH2-MgCl
Concentration 1.2 to 1.5M
Fieser 16,118; 15,343; 12,540; 06,636; 05,724; 10,433
Density 1.0400g/mL
Solubility Information Solubility in water: reacts violently with water.
IUPAC Name [(chloromagnesio)methyl]trimethylsilane
Formula Weight 146.97
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.04
Quantity 100 mL
Flash Point −17°C
Physical Form Low Melting Solid
Chemical Name or Material (Trimethylsilyl)methylmagnesium chloride, 1.1M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Trimethylsilyl)methylmagnesium chloride, 1.1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'