missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10254824
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
CAS 1066-45-1, 110-54-3
Molecular Formula C3H11ClSn
MDL Number MFCD00000520
Concentration 1M (NMR), exact strength on the certificate of analysis
Density 0.7970g/mL
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
SMILES Cl.C[SnH](C)C
IUPAC Name chloro(trimethyl)stannane
Molecular Weight (g/mol) 201.28
PubChem CID 14016
Formula Weight 199.25
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.797
Quantity 100 mL
Flash Point −23°C
Physical Form Liquid
Chemical Name or Material Trimethyltin chloride, 1M (25 wt.%) solution in Hexanes
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethyltin chloride, 1M (25 wt.%) solution in hexanes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'