missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/2D Barcode

Product Code. p-10316836 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
55mm contact plates
90 mm monoplate
Fill Weight:
13 g
30 g
Includes:
VHP Indicator, Neutralisers
VHP Indicator, no Neutralisers
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Fill Weight Includes unitSize
30042332 55mm contact plates 13 g VHP Indicator, Neutralisers Pack of 100
30042333 90 mm monoplate 30 g VHP Indicator, Neutralisers Pack of 100
30042334 90 mm monoplate 30 g VHP Indicator, no Neutralisers Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30042332 Supplier Thermo Scientific™ Supplier No. PO4011D

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Experience next-level cleanroom confidence with these plates, complete with VHP indicator, lecithin, polysorbate 80, sodium thiosulphate, L-histidine and 2D barcode.

Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar Plates with VHP Indicator and 2D Barcode.

Achieve confidence in your results and ensure optimum media growth using plates with a vapor-phase hydrogen peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:

  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover

Conveniently sold in packs of 100 which are sealed for security
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture.

Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Format 55mm contact plates
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 13 g
Includes VHP Indicator, Neutralisers
Packaging Type Triple Wrapped - Irradiated
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.