missing translation for 'onlineSavingsMsg'
Learn More

Tripropylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-3737516
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
102-69-2
C9H21N
143.27
MFCD00009363
YFTHZRPMJXBUME-UHFFFAOYSA-N
tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
7616
CHEBI:38880
N,N-dipropylpropan-1-amine
CCCN(CCC)CCC
This item is not returnable. View return policy
102-69-2
C9H21N
143.27
MFCD00009363
YFTHZRPMJXBUME-UHFFFAOYSA-N
tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
7616
CHEBI:38880
N,N-dipropylpropan-1-amine
CCCN(CCC)CCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -93.0°C
Color Colorless
Density 0.7600g/mL
Boiling Point 156.0°C
Flash Point 32.5°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. dipropylamine (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2)3N
Refractive Index 1.4155 to 1.4175
Quantity 1 L
Beilstein 04, II, 623
Specific Gravity 0.76
Solubility Information Solubility in water: 2.6g/L (20°C). Other solubilities: soluble in most organic solvents
Viscosity 0.627 mPa.s (25°C)
Formula Weight 143.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tripropylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'