missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tris(1-methoxy-2-methyl-2-propoxy)bismuth is used as metal organic precursors to prepare bismuth titanium oxide thin films, which finds application in ferroelectric random access memory.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Melting Point | 70°C |
Color | Yellow |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethylcyclohexane. |
Formula Weight | 518.4 |
Percent Purity | 99.99% |
Physical Form | Crystalline |
Assay | (metals basis) |
Chemical Name or Material | Tris (1-methoxy-2-methyl-2-propoxy)bismuth |
Safety and Handling
Product Identifier
- Tris(1-methoxy-2-methyl-2-propoxy)bismuth
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'