Learn More
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
Brand: Fisher Bioreagents BP1338-1
49.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 32
Additional Details : CAS Number : 38641-82-6 Weight : 1.05000kg
Description
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology
- Simplify your buffer prepraration by using ready-made Fisher BioReagents 100X concentrate TE buffer solution -simply dilute as needed
- 100X solution contains 1.0M tris and 0.1M EDTA
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer (pH 8.0) is routinely used for resuspending DNA after precipitation
- Intended for laboratory research use only
Specifications
Tris-EDTA | |
77-86-1,60-00-4,7732-18-5 | |
Liquid | |
DNase free | |
Nalgene Poly Bottle | |
TE |
100X Solution | |
Undesignated | |
1 L | |
Filtered through a 0.2-micron filter. | |
Protease free | |
Molecular Biology |