Learn More
Tris(hydroxymethyl)aminomethane, Merck Millipore
Catalog Number 10001240
Click to view available options
100 g
GR for analysis buffer substance ACS (Reag. Ph Eur)
  • Biological buffer substance
  • Tris is widely used as a biological buffer substance in the pH range of 7-9.
  • For example it is used for the freezing of proteins.
  • The grade conforms precisely with the Reag. Ph. Eur. requirements.
  • Application:
    • Tris(hydroxymethyl)aminomethane GR for analysis buffer substance ACS, Reag. Ph Eur. CAS 77-86-1, EC 201-064-4, chemical formula H2NC(CH2OH)3