missing translation for 'onlineSavingsMsg'
Learn More

TTC21A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA557301

Additional Details : Weight : 0.02000kg

Product Code. 15725201

  • 530.00 € / 100µL
Estimated Shipment: 11-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: QGAESNYMEK KELEQQGVST AEKLLREFYP HSDSSQTQLR LLQGLCRLAT REKANMEAAL GSFIQIAQAE KDSVPALLAL AQAYVFLKQI PK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

TTC21A Polyclonal antibody specifically detects TTC21A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TTC21A
Polyclonal
Unconjugated
TTC21A
IFT139A; STI2; Stress-inducible protein 2; testicular tissue protein Li 212; tetratricopeptide repeat domain 21A; tetratricopeptide repeat protein 21A; TPR domain containing STI2; TPR repeat protein 21A; TTC21A
Rabbit
Antigen affinity chromatography
RUO
199223
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NDW8
TTC21A
Recombinant protein corresponding to Human TTC21A
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only