Learn More
Gosselin™ Tube Closures
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
Brand:  Gosselin™ BA110-25
Code : A0
Additional Details : Weight : 1.42200kg
 View more versions of this product
Product Code. 11748603
  • 66.00 € / Pack of 2500
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
Specifications
Specifications
Ribbed Closure Plug
LDPE
Yellow
2500 Bag