Learn More
Scientific Industries SI™ Tube Rack
Accommodates up to 10 tubes of any material. up to 29mm diameter. Scientific Industries SI™ Tube Rack is for use with Vortex-Genie™ MAX.
157.00 € - 309.00 €
Product Code | Brand | No. of Positions | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Packaging Quantity | Price | Quantity & Availability | ||||
15537355
|
Scientific Industries SI™
SI-MX010 |
10 | 1 Ea. |
157.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15547355
|
Scientific Industries SI™
SI-MX020 |
20 | 2/Pk. |
309.00 €
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
Description
- Holds up to 10 tubes
- For use with Vortex-Genie™ MAX
Specifications
29 mm dia. Tubes |