missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tungsten wire basket; # turns, 9; ID(mm), 4; Ht(mm), 7; Wire leads(cm), 3.5; Wire dia(mm) 0.51

Brand:  Alfa Aesar™ 041177.KK

Code : NEW

Additional Details : CAS Number : 7440-33-7 Weight : 0.01000kg

Quantity 10 pc.

 View more versions of this product

Product Code. 15414855

  • 67.10 € / Pack of 10


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
CHEBI:27998
[W]
W
MFCD00011461
23964
tungsten

Specifications

Tungsten wire basket
Turns: 9, ID: (4mm), Ht(7mm), Wire leads (3.5cm), Wire dia (0.51mm)
W
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire
10 pc.
7440-33-7
MFCD00011461
[W]
183.84
CHEBI:27998
Videos
SDS