Learn More
Thermo Scientific™ TurboFect in vivo Transfection Reagent
Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.
Brand: Thermo Scientific™ R0541
Additional Details : Weight : 0.00900kg
Description
TurboFect Transfection Reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect is a sterile solution of a proprietary cationic polymer in water. The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.
Highlights
- High transfection efficiency of plasmid DNA into a wide variety of cell types
- Adherent and suspension cells
- Differentiated and undifferentiated cells
- Excellent transfection efficiency in the presence or absence of serum
- Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
- Simple protocol
Review our experimental considerations to improve your plasmid transfection results.
Review our experimental considerations to improve your plasmid transfection results.
Specifications
Animal Origin-Free, Chemically Defined | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Chemical Transfection | |
TurboFect | |
120 μL |
Cells in Vivo | |
Plasmid DNA | |
Transfection | |
Transfection Reagent |