missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TWEAK R (CD266/TNFRSF12A) Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMTNFRSF12A

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15415343

  • 620.00 € / 1 set
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Mouse TWEAK R (CD266/TNFRSF12A) ELISA quantitates Ms TWEAK R in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms TWEAK R. Principle of the method The Mouse TWEAK R solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by biding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in aorta and liver. It may also play a role in vascular cell migration and angiogenesis.
Specifications

Specifications

Q9CR75
95 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
27279
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Angiogenesis
2°C to 8°C
Mouse
4 hr. 45 min.
102.2-25,000 pg/mL
95pg/mL to 25000pg/mL
Biotin
Chemokine & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, TWEAK, VCAM-1, VEGF.
Mouse
Colorimetric Microplate Reader
AI255180,C87282,Fn14,HPIP,TWEAK-R,TweakR
<10%
HRP
RUO
Plasma, 33 μL; Serum, 33 μL; Supernatant, 100 μL
AI255180, C87282, Fn14, HPIP, TWEAK-R, TweakR
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers