missing translation for 'onlineSavingsMsg'
Learn More

Tween™ 80, Fisher BioReagents

Catalog Number 10592955
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Bottle
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Non-ionic detergent for selective protein extraction

This nonionic detergent is used in the isolation of water-soluble membrane proteins.
TRUSTED_SUSTAINABILITY
Color Amber
CAS 9005-65-6
Physical Form Liquid
Density 1.080g/cm³
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL Number MFCD00082107
pH 6
Quantity 500 mL
Merck Index 15, 7703
Synonym Polysorbate 80, Polyoxyethylene-20-sorbitan Monooleate
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Hydroxyl Value 65 to 80
Molecular Weight (g/mol) 604.82
Identification Pass Test
Ignition Residue 0.25% max.
Packaging HDPE Bottle
Water 3% max.
Chemical Name or Material Tween™ 80
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'