missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TX-750 Round Bucket Adapters

Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.

Brand:  Thermo Scientific™ 75003710

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10012759

  • 677.15 € / Pack of 4
Estimated Shipment: 21-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.

Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications

Specifications

250mL Bottle
250mL Bottle
Round Bucket Adapter
250mL
250 mL Nalgene Bottle, 225 mL/175 mL BD Falcon Conical Bottle (requires BD #352090) (Open buckets only), 200 mL Nunc Conical Bottle (requires Nunc #377585) (Open buckets only), 175 mL Nalgene Conical Bottle (requires Nalgene #DS3126-0175) (Open buckets only)
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers