missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 S
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Seam Type:
Stitched
Stitched, Taped
Size (Metric) Chest:
100 to 108 cm
108 to 116 cm
116 to 124 cm
124 to 132 cm
84 to 92 cm
92 to 100 cm
Description
- DuPont™ Tychem™ 4000 S. Hooded coverall available in white, in sizes SM to 3X. Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up. Self-adhesive chin flap for tight seal of suit to face-mask.
Tychem™ 4000 garments are made from a lightweight, supple and durable fabric (<700g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek™ substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
Tychem™ 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services.
Specifications
Product Type | Chemical Protective Clothing |
Size | Small |
Gender | Unisex |
Material | Tychem |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Closure Type | Double Zipper Front with Double Flaps |
Seam Type | Stitched, Taped |
Sterility | Non-sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'