missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-PEGASUS-90 Single Door
Safe and approved storage of flammable hazardous substances in working areas
2838.15 € - 3468.85 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 615 mm |
Depth (Metric) Interior | 522 mm |
Dimensions (D x W x H) | 615 x 599 x 1953 mm |
Dimensions (D x W x H) Interior | 522 x 450 x 1647 mm |
Product Code | Brand | Configuration | Door Style | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Door Style | Description | Price | Quantity & Availability | |||
15991906
|
asecos™
3007404133600 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Light gray doors |
2838.15 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15942196
|
asecos™
3007504134371 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Light gray doors |
2838.15 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15922156
|
asecos™
3007504034371 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Warning Yellow doors |
2838.15 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15911886
|
asecos™
3007404033606 |
3 x tray shelf, 1 x perforated insert (180 mm high), 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Warning Yellow doors |
3039.55 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15962156
|
asecos™
3007504034375 |
3 x tray shelf, 1 x perforated insert (180 mm high), 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Warning Yellow doors |
3039.55 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15941916
|
asecos™
3007404133608 |
4 x 5 liter drawer (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Light gray doors |
3189.01 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15921886
|
asecos™
3007404033608 |
4 x 5 liter drawer (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Warning Yellow doors |
3189.01 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15982196
|
asecos™
3007504134376 |
4 x 5 liter drawer (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Light gray doors |
3189.01 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15972156
|
asecos™
3007504034376 |
4 x 5 liter drawer (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Warning Yellow doors |
3189.01 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15961916
|
asecos™
3007404133610 |
6 x 5 liter drawer (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Light gray doors |
3468.85 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15941886
|
asecos™
3007404033610 |
6 x 5 liter drawer (sheet steel powder-coated) | Swing, Left-hinged, Auto-close | Warning Yellow doors |
3468.85 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15902206
|
asecos™
3007504134378 |
6 x 5 liter drawer (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Light gray doors |
3468.85 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15992156
|
asecos™
3007504034378 |
6 x 5 liter drawer (sheet steel powder-coated) | Swing, Right-hinged, Auto-close | Warning Yellow doors |
3468.85 €
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
522 mm | |
522 x 450 x 1647 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
Powder-coated textured | |
450 mm |
615 mm | |
615 x 599 x 1953 mm | |
90 mm | |
Safety storage cabinet | |
1953 mm | |
1 | |
599 mm |