missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Touchless-90 Warning Yellow
Safe and approved storage of flammable hazardous substances in working areas
3254.20 € - 5172.80 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
---|---|
Material | Steel |
Product Type | Safety storage cabinet |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Height (Metric) Exterior | 1968 mm |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Depth (Metric) Exterior | Configuration | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Depth (Metric) Exterior | Configuration | Price | Quantity & Availability | |||
15953766
|
asecos™
3023700233659 |
596 mm | 616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15993766
|
asecos™
3023700233663 |
596 mm | 616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15903776
|
asecos™
3023700233681 |
596 mm | 616 mm | 4 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15923776
|
asecos™
3023700233683 |
596 mm | 616 mm | 6 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15934276
|
asecos™
3031700233695 |
596 mm | 749 mm | 4 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15954276
|
asecos™
3031700233697 |
596 mm | 749 mm | 6 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15922756
|
asecos™
3012000233715 |
1196 mm | 616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15962756
|
asecos™
3012000233719 |
1196 mm | 616 mm | 3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15912766
|
asecos™
3012000233724 |
1196 mm | 616 mm | 4 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15932766
|
asecos™
3012000233726 |
1196 mm | 616 mm | 6 x drawer (sheet steel powder-coated) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Effortless use: the sensor detects a person within approx. 2 seconds
- User-friendly and safe: after detection, the folding doors open automatically within approx. 5 seconds. The sensor recognises the person while operating and hold the folding doors open
- Always closed: if the person leaves the sensor-controlled detection area, the folding doors close automatically after approx. 30 seconds
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
Safety storage cabinet | |
1968 mm | |
Powder-coated plain |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm |