missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet XL-CLASSIC-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3039300130395
Additional Details : Weight : 837.06842kg
Alert:
For indoor use only
Description
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with cylinder lock
- Internal flexibility: transport base (optional) for safe and easy in-house transport
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Safety while decanting: end-to-end earthing link at the rear panel, inside with earthing wires incl. clamp
- Certificates
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- GS certified
- CE compliant
Specifications
Safety storage cabinet | |
Light Gray | |
1105 mm | |
Powder-coated plain | |
XL90.222.110.WDAS | |
1890 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1018 x 1105 x 2224 mm | |
864 mm | |
1 x transport base (steel powder-coated), 1 x bottom collecting sump with grid (sheet steel galvanized) |
Steel | |
936 mm | |
918.50 kg | |
2 | |
Door open arrest system | |
2224 mm | |
Wing | |
864 x 936 x 1890 mm | |
1018 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, GS certified, CE compliant |