missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Type A Safety Bumper
Protect corners and edges with this Type A Safety Bumper. Dual function: indicates hazardous situations and softens any impacts.
Brand: Brady™ 225022
Code : NEW
Additional Details : Weight : 0.47200kg
Description
- Highly resistant against hydrocarbons.
Specifications
10 Pk. | |
Self-adhesive Safety Bumpers - Corner (a) | |
Safety Identification | |
Safety Bumpers | |
Cellular Thermoplastic |
26.77 in. | |
Black and Yellow Striped | |
110 mm | |
4.33 in. | |
0.68 m |