missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Type C Safety Bumper
Protect corners and edges with this Type C Safety Bumper. Dual function: indicates hazardous situations and softens any impacts.
Brand: Brady™ 225024
Code : NEW
Additional Details : Weight : 1.75000kg
Description
- Highly resistant against hydrocarbons.
Specifications
10 Pk. | |
Self-adhesive Safety Bumpers - Cover (c) | |
Safety Identification | |
Safety Bumpers | |
Cellular Thermoplastic |
29.53 in. | |
Yellow | |
151 mm | |
5.94 in. | |
0.75 m |