missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3758966
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
60-19-5
C8H12ClNO
173.64
MFCD00012901
RNISDHSYKZAWOK-UHFFFAOYSA-N
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
66449
[H+].[Cl-].NCCC1=CC=C(O)C=C1
This item is not returnable. View return policy
60-19-5
C8H12ClNO
173.64
MFCD00012901
RNISDHSYKZAWOK-UHFFFAOYSA-N
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
66449
[H+].[Cl-].NCCC1=CC=C(O)C=C1

Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Tyramine is a naturally occurring monoamine that is derived from the decarboxylation of the amino acid tyrosine utilized in human and invertebrate nervous systems
• Tyramine is synthesized and released from neurons and acts as a neuromodulator upon binding to tyramine receptors
• Tyramine has been found to impact the immune system and cardiovascular system in humans
Applications
• Tyramine may be used to form tyramide conjugate, which can be used as a signal amplification reagent in immunofluorescence assays
• Tyramine-hyaluronic acid conjugates can be used to form biodegradable hydrogels, with utility in drug delivery and tissue engineering
• Tyramine is useful for laboratory studies of neuronal stimulation
TRUSTED_SUSTAINABILITY
Melting Point 271°C to 274°C
Color White
Boiling Point 269°C
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2NH2·HCl
Quantity 5 g
Beilstein 13, 625
Merck Index 15, 10016
Solubility Information (10% in water) Clear colorless to light yellow
IUPAC Name hydrogen 4-(2-aminoethyl)phenol chloride
Formula Weight 173.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tyramine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'