Learn More
Corning™ Universal Fit Hinged Rack Pipet Tips, Sterile
Hinged closure provides easy stacking and steam penetration during autoclaving
Brand: Corning™ 4714
Additional Details : Weight : 2.60930kg
Description
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
100 to 1000 μL | |
Blue | |
Hinged Rack | |
Hinged rack |
Sterile | |
Hinged closure provides easy stacking and steam penetration during autoclaving | |
Round | |
Yes |