missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Inlet Solvent Filters
Stainless-steel or multiple materials
25.20 € - 189.00 €
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
12943160
|
Idex
42711-03 |
10 μm | 5/Pk. |
189.00 €
Pack of 5 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
12963160
|
Idex
42711-05 |
2 μm | 5/Pk. |
125.00 €
Pack of 5 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
12936239
|
Kinesis™
42711-07 |
20 μm | 1/pk. |
84.30 €
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
12953160
|
Idex
42711-04 |
2 μm | 1/pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Cups and end fittings sold separately, see 05-704-54 and 045-707-83.
Specifications
Inlet Solvent Filter |