missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Inlet Solvent Filters: End Fittings

Manufactured from Polyether ether ketone (PEEK). Upchurch Scientific Inlet Solvent Filters: End Fittings are used for Analytical HPLC Systems.

Brand:  Kinesis™ 02010-08

Code : Z3

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 12946289

  • 165.00 € / Pack of 10
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • For Analytical HPLC System
  • 0.5μm PEEK filter end fittings
  • Black body
  • Swept volume of 5.7μL
  • Thread port: 10-32 Coned

  • For Analytical HPLC System
  • 0.5μm PEEK filter end fittings
  • Black PEEK body
  • Swept volume of 5.7μL
Specifications

Specifications

0.5 μm
Replacement Filter End Fitting
PEEK
10 to 32 Coned
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers