missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Upright ULT Freezer Sliding Tray Racks

Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.

Brand:  Fisherbrand™

Additional Details : Weight : 2.92000kg

 View more versions of this product

Product Code. 15759559

  • 175.00 € / Each
Estimated Shipment: 28-08-2024
to see stock.
Add to basket

Description

Description

  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Sliding trays provide more flexibility and easy access to the boxes
Specifications

Specifications

Sliding Tray Rack
Stainless Steel
Upright ULT Freezer
12 (2in.) boxes
Standard 2 in. Cryoboxes
419 x 140 x 225 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers