Learn More
USB Communication Cable for Eutech™ Temp 300 Series meters
Eutech™ provides various types of cables and adapters available for use with Eutech™ different meters.
Brand: Thermo Scientific Eutech 30X379302
Code : 04
Additional Details : Weight : 0.10000kg
Description
Eutech™ provides various types of cables and adapters available for use with Eutech™ different meters. The USB Communication Cable is used for Eutech™ Temp 300/Temp 360 thermocouple datalogger.
- USB to USB-mini cable available for Eutech Temp 300 / Temp 360 meters.
Specifications
USB cable | |
USB Communication Cable |