Learn More
OHAUS™ USB Interface Kit
For use with Ranger 3000, Ranger Count 3000 compact counting scales and Valor 7000 scale
Brand: OHAUS™ 30037449
Code : NEW
Additional Details : Weight : 0.25000kg
Includes: USB PC board, cable, CD, pressure button, four screws, instruction manual
Description
Unique solution to the problem of connecting a scale to a computer using a USB
System Requirements
- PC running Windows 98™ , Windows 98SE ™ , Windows ME ™ , Windows 2000 ™ , Windows XP ™ or Windows 7 ™
- Available USB port
Specifications
Ranger 3000, Ranger Count 3000 Compact Counting Scales | |
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
USB Interface Kit |
USB PC board, cable, CD, pressure button, four scr | |
Interface Cables and Kits |