Learn More
Uvex™ Safety Glasses Skyguard NT
17.87 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Product Type:
Safety Glasses Skyguard NT UVEX
Description
- Soft seal conforms to EN166 1F 34 KN offering protection against liquid droplets, splashes and large dust particles
- Injection moulded TPU frame design significantly increase swearer protection by minimising the risk of particle ingress due to
- Hard and soft high tech materials are combined creating soft effective seal around the eyes, reduced weight and is easy to clean
- Temple length adjustment enables the wearer to achieve an individual fit
- Low profile with closer fitting, smaller dimensions for high levels of protection and is less obtrusive for increased personal movement
- Wraparound design for an unhindered field of vision
- Permanent uvex supravision lens coating, permanent anti-fog coating on the inside of the lens, scratch resistant on the outside
Specifications
Product Type | Safety Glasses Skyguard NT UVEX |