Learn More
Buerkle™ V4A Stainless Steel PharmaScoop
Buerkle™ V4A Stainless Steel PharmaScoop™ is made without edges, crevices or undercuts, to avoid cross-contamination.
86.60 € - 292.00 €
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | ||||
15578831
|
Buerkle™
5324-0010 |
10 mL | 25 x 45 mm |
86.60 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
15588831
|
Buerkle™
5324-0011 |
50 mL | 38 x 70 mm |
147.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
10263652
|
Buerkle™
5324-0016 |
50 mL | 38 x 70 mm |
245.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10355522
|
Buerkle™
5324-0012 |
100 mL | 100 x 50 mm |
200.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
11911561
|
Buerkle™
5324-0018 |
500 mL | 140 x 95 mm |
201.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10435762
|
Buerkle™
5324-0013 |
750 mL | 210 x 95 mm |
206.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
10204642
|
Buerkle™
5324-0014 |
1100 mL | 235 x 110 mm |
229.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10545904
|
Buerkle™
5324-0015 |
1250 mL | 195 x 125 mm |
236.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
15229058
|
Buerkle™
5324-0017 |
2500 mL | 215 x 185 mm |
292.00 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
Description
- Stainless steel AISI 316 (1.4401)
- GMP conform
- Highly polished
- Highly polished inner and outer surface