Learn More
Wheaton™ 47 mm Filtration Assemblies with Stainless Steel Support
Designed for filtering viscous or proteinaceous solutions or to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assemblies with Stainless Steel Support features glass components manufactured from low extractable borosilicate glass.
Brand: Wheaton™ 419360
284.23 EUR valid until 2024-12-31
Use promo code "21897" to get your promotional price.
Code : A0
Additional Details : Weight : 0.90720kg
Description
- Glass conforms to ASTM E 438 Type I, Class A requirements
- Includes a stainless steel support with 120 mesh screen, PTFE support screen gasket, support base, 300mL graduated funnel, anodized clamp and a No. 8 silicone stopper
Specifications
Analysis Of Particulate Or Microbiological Contamination | |
Vaccum Filter | |
1000 mL |