Learn More
BD Vacutainer™ ESR Seditainer™ Tubes
Designed for ESR determinations without needing to open the tube. BD Vacutainer™ ESR Seditainer™ Tubes are 5mL glass tubes containing buffered sodium citrate for use with the BD Seditainer™ Manual Stand. Tubes are available with a black Hemogard™ or conventional closure.
Brand: BD 366671
Code : 02
Additional Details : Weight : 3.50000kg
Description
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. BD Seditainer™ tubes are designed for ESR determination without having to use a sedimentation pipette.- Whole blood is drawn directly into the evacuated glass tube
- Tubes are mixed by inverting the tube 180° and back, 8-10 times
- Up to 10 tubes are placed directly into the BD Seditainer™ Manual Stand
- There is no need to remove the tube stopper and pipette or transfer the sample
- Results can be read after 60 minutes; results correspond to the Westergren method
- BD Seditainer™ Manual Stand holds up to 10 tubes and has a height adjustable zero mark
Specifications
Evacuated Blood Collection Tube | |
Glass | |
Hemogard | |
BD Seditainer™ Manual Stand; BD Vacutainer™ Needles and Tube Holders | |
10.25 x 120 mm | |
Block |
Sodium Citrate (0.105M) | |
Sterile | |
5 mL | |
Black | |
ESR Determination |