missing translation for 'onlineSavingsMsg'
Learn More

Edwards Vacuum Rotary Vane Vacuum Pump Exhaust Oil Mist Filters

Captures oil mist from pump outlet when used with RV Rotary Vane Pumps

Brand:  Edwards A46229000

Code : 03

Additional Details : Weight : 30.00000kg

 View more versions of this product

Product Code. 10034970

  • 603.59 € / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Includes: Supplied with NW25 clamp, centering ring, “O” ring, and NW25 to 3/4 in. adapter.

Description

Description

Captures oil mist from pump outlet when used with RV Rotary Vane Pumps

Ejection of oil into the atmosphere during usage of gas ballast or pump high gas throughputs can be prevented.

  • Semi-transparent white bottom half for oil level monitoring
  • Integral pressure relief valve opens if oil element is blocked
  • Neutralize oil mist smell
  • Integral pressure relief valve opens if oil element blocks
  • Efficient at 99.999% DOP test
  • Azide proof

Use optional oil return kits for reduce oil level maintenance and savings in pump oil

Certifications

Meets COSHH and health and safety regulations

Specifications

Specifications

Model RV12, E2M18 pump
NW25 clamp, centering ring, O Ring, NW25 to 0.75 in. BSP adapter
Oil Mist Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers