Learn More
BUCHI Vacuum Seals for Rotary Evaporators
Ensure efficient operation of your Rotavapor by replacing the vacuum seal as needed. Designed specifically for BUCHI Rotavapors and glassware.
Brand:  BUCHI 10179
Code : A0
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 10499302
  • 127.00 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • High chemical resistance
  • OEM seal for proper fit and performance
Specifications
Specifications
Gasket with Ring
Rotavapor R-1X4, R-3