Learn More
Emma Safety Footwear Valentino (Frontier 164) Safety Shoes
Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
Brand:  Emma Safety Footwear MM16409045
Additional Details : Weight : 2.24700kg
 View more versions of this product
Product Code. 15958281
  • 222.60 € / 1 pair
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • TPU Outsole
  • Full grain leather
  • ESD Certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
45
Laces
Valentino (Frontier 164)
Leather, Nubuck
Steel