Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Brand:  Thermo Scientific™ 699-119300
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 11947471
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Packages provide:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification
Specifications
Specifications
Validator Lite for Evolution 60S, BioMate 3S, GENESYS 10S UV-Vis and GENESYS 10S UV-Vis Bio
Evolution 60S/60; BioMate 3S; GENESYS 10S UV-Vis Bio/10S UV-Vis
UV Validator Lite