missing translation for 'onlineSavingsMsg'
Learn More

Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 15416445
Click to view available options
Quantity:
25 Pc.
5 Pc.
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 Pc.
Percent Purity 99.8%
Odor Odorless
Weight ≈2.3g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Vanadium slug, 11mm (0.43 in.) dia. x 4mm (0.16 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'