missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Vancomycin hydrochloride is a glycopeptide antibiotic and antiviral from Streptomyces orientalis
- It shows antibiotic activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains
- Vancomycin hydrochloride inhibits bacterial mucopeptide synthesis
Applications
- Vancomycin hydrochloride demonstrates potent activity against gram-positive bacteria
- It has been shown in studies to be highly effective against methicillin-resistant Staphylococcus aureus (MRSA) in vitro and in vivo
Specifications
Type | Vancomycin Hydrochloride |
Physical Form | Powder |
pH | 2.5 to 4.5 (5% soln.) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: 50g/L. Other solubilities: moderately soluble in diluted methanol, insoluble in higher alcohols, acetone and ether |
Formula Weight | 1485.73 |
Percent Purity | ≥80.0% (vancomycin B) (HPLC), 9.0% (individual impurities) max., greater than 900μg/mg (potency, on anhydrous substance) |
Water | 5% max. |
Color | Tan to White |
Show More |
Safety and Handling
Product Identifier
- Vancomycin hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'