missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vancomycin Hydrochloride MP Biomedicals
Click to view available options
Quantity:
250 mg
Specifications
Type | Vancomycin Hydrochloride |
CAS | 123409-00-7 |
Molecular Formula | C66H257Cl2N9O24 |
InChI Key | ARNOCQCQWLXLRG-UHFFFAOYSA-N |
SMILES | C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl |
IUPAC Name | azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride |
Molecular Weight (g/mol) | 1632.721 |
PubChem CID | 44134997 |
Quantity | 250 mg |
Chemical Name or Material | Vancomycin Hydrochloride |
Safety and Handling
Product Identifier
- Vancomycin Hydrochloride
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : vancomycin hydrochloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'