Learn More
VARS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564437
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NALRFILNAL GEKFVPQPAE ELSPSSPMDA WILSRLALAA QECERGFLTR ELSLVTHALH HFWLHNLCDV YLE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
VARS2 Polyclonal antibody specifically detects VARS2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
VARS2 | |
Polyclonal | |
Unconjugated | |
VARS2 | |
1190004I24Rik; COXPD20; Kiaa1885; mKIAA1885; valine tRNA ligase 2, mitochondrial (putative); valine--tRNA ligase, mitochondrial; ValRS; valyl trna synthetase; Valyl-tRNA synthetase; valyl-tRNA synthetase 2, mitochondrial; valyl-tRNA synthetase 2, mitochondrial (putative); valyl-tRNA synthetase like; valyl-tRNA synthetase, mitochondrial; Valyl-tRNA synthetase-like; VARS2; VARS2L; VARSL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5ST30 | |
VARS2 | |
Recombinant protein corresponding to Human VARS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only