Learn More
Emma Safety Footwear Vera XD Safety Shoes, Red
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
Brand: Emma Safety Footwear MM50752445
Additional Details : Weight : 1.46100kg
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Duo Heat Resistant Outsole
- Chrome-free lining
- Sanitized
Specifications
Vera XD, Red | |
Microfiber | |
45 |