Learn More
3M™ Versaflo™ M-400 Series Respiratory Helmet Assembly
Used with an approved air source, M-400 Series Helmets provide respiratory (OSHA APF=1000) and limited head (ANSI Z89.1-2003 Type 1 Class G) and eye (ANSI Z87.1-2010) protection.
Brand: 3M™ 7000104037
568.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : S8
Additional Details : Weight : 0.01000kg
Description
Features:
- Chemical and Scratch Resistant Lenses
- Large Viewing Area
- Lightweight, Easy Visor Replacement System
Environmental Conditions:
- Cold Working Environment
- High Mobility Required
- Safety Helmet Required
Hazard Type:
- Gas, Vapour and Particulates
- Liquid Splash
- Low Energy Flying Particles/Dust
- Falling Objects
- Sparks and Hot Particles
Includes:
- The M-405 assembly includes a M-925 Visor, M-444 Inner Collar, and M-445 Outer Shroud.
- The M-925 visor is made of polycarbonate.
- The M-445 shroud is appropriate for general purpose use.
- The M-407 Assembly includes a M-927 Premium Visor, M-444 Inner Collar, and M-447 Flame Resistant Outer Shroud.
- The M-927 visor is made of coated polycarbonate.
- The M-447 shroud is made of Nomex flame resistant fibers.
- 3M Catalog# M-405 , 3M Catalog# M-407
Specifications
NPF: 200 (V-Series), NPF: 500 (Jupiter/TR-302E/TR-602E) | |
Black | |
Respirator Helmet | |
3M Air Delivery Units | |
Polycarbonate | |
Non-sterile | |
Polycarbonate | |
346 x 346 x 254 mm | |
346 mm | |
346 mm | |
254 mm | |
1066 g |
CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397 | |
Nylon | |
Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction | |
Highly Durable Shroud | |
M-406 | |
Textile | |
No | |
13.6 in. | |
13.6 in. | |
9.6 in. | |
2.4 lb. |