Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 800 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
Brand:  Cytiva (formerly Pall Lab) 66401
Additional Details : Weight : 0.26600kg
 View more versions of this product
Product Code. 17184561
  • 219.00 € / Pack of 100
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Versapor (Acrylic Copolymer on a Non-woven Support)
Non-sterile
47 mm
0.8 μm
94 μm