missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits
Shop All Macherey-Nagel ProductsDescription
- Colorimetric and titrimetric test kits
- Avoids hazardous substances wherever possible
Specifications
Specifications
| For Use With (Application) | To determine ammonia in water |
| Includes | 30mL NO3-, 5 g NO3-, 70mm measuring spoon, 2 screw-plug measuring glasses, slide comparator, color chart, 5mL plastic syringe, Instructions for use |
| Range | 0 · 1 · 3 · 5 · 10 · 20 · 30 · 50 · 70 · 90 · 120 mg/l NO3- |
| Sufficient For | 110 Tests |
| Test Method | Colorimetric method |
| Type | Visocolor Eco Nitrate |
Safety and Handling
Product Identifier
- VISOCOLOUR ECO nitrate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- MIXTURE LIST-Contain: zinc powder (stabilized)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?