missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ ECO Nitrite Colorimetric Test Kit

Product Code. 10010442 Shop All Macherey-Nagel Products
Change view
Click to view available options
:
For Use With: Colorimetric determination of nitrite
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10010442 For Use With: Colorimetric determination of nitrite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010442 Supplier Macherey-Nagel™ Supplier No. 931244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) Colorimetric Determination of Nitrate
compliance-icons
Product Identifier
  • VISOCOLOUR ECO nitrite, refill pack
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Supplemental information
  • MIXTURE LIST-Contain: sulphanilamide, o-phosphoric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.