missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Ejector Sleeves

For adjustable- and fixed-volume pipetters

Brand:  VistaLab™ Technologies 9057-3011

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11936032

  • 18.38 € / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • ESC: 0.5-20μL, 2-125μL & 5-250μL
  • QS & M: 02.-2μL, 1-10μL, 2-20μL, 10-100μL, 20-200μL
  • F1 & F2: all models dispensing 20oμL and less
Specifications

Specifications

Electronic Single Channel Model 25 to 1250 μL, Adjustable Model 100 to 1000 μL, All Fixed Volume Models Dispensing 201 μL and Above
5 in.
1 in.
1 Ea.
3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions