missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Pipette Tips: Low Retention
For use with Ovation and MLA Brand pipetters
87.40 € - 238.00 €
Specifications
Color | Clear |
---|---|
Rack Color | Blue |
Thickness | 5 in. |
For Use With (Application) | Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications |
Format | Rack |
Product Code | Brand | Volume (Metric) | Sterility | Filtered | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Filtered | Price | Quantity & Availability | |||
11915992
|
VistaLab™ Technologies
4070-1333LR |
2 μL | Sterile | Filtered |
238.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11955992
|
VistaLab™ Technologies
4070-3004LR |
1250 μL | Non-sterile | Non-filtered |
137.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11995992
|
VistaLab™ Technologies
4070-1002LR |
25 μL | Non-sterile | Non-filtered |
91.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11975992
|
VistaLab™ Technologies
4070-3132LR |
1250 μL | Sterile | Non-filtered |
149.00 €
Pack of 960 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
11905992
|
VistaLab™ Technologies
4070-3332LR |
1250 μL | Sterile | Filtered |
196.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11906002
|
VistaLab™ Technologies
4070-1032LR |
25 μL | Sterile | Non-filtered |
122.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11945992
|
VistaLab™ Technologies
4070-2004LR |
250 μL | Non-sterile | Non-filtered |
87.40 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11935992
|
VistaLab™ Technologies
4070-2132LR |
250 μL | Sterile | Non-filtered |
122.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11925992
|
VistaLab™ Technologies
4070-1332LR |
25 μL | Sterile | Filtered |
191.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11965992
|
VistaLab™ Technologies
4070-2332LR |
250 μL | Sterile | Filtered |
182.00 €
Pack of 960 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
Description
- Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
- Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Clear | |
5 in. | |
Rack | |
192 Tips/Rack | |
No |
Blue | |
Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications | |
Low Retention, Straight | |
Single-use |